Skip to Content
Merck
All Photos(1)

Key Documents

PHR1768

Supelco

Dexamethasone Sodium Phosphate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Dexamethasone 21-phosphate disodium salt, 9-Fluoro-11b,17, 21-trihydroxy-16α- methylpregna-1,4-diene-3,20-dione 21-(dihydrogen phosphate) disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€138.00

€138.00


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€138.00

About This Item

Empirical Formula (Hill Notation):
C22H28FNa2O8P
CAS Number:
Molecular Weight:
516.40
Beilstein:
6473066
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€138.00


Estimated to ship on01 April 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 108
traceable to Ph. Eur. D0720000
traceable to USP 1177032

API family

dexamethasone

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@@H](C)[C@]4(O)C(=O)COP([O-])([O-])=O

InChI

1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15+,16+,17+,19+,20+,21+,22+;;/m1../s1

InChI key

PLCQGRYPOISRTQ-FCJDYXGNSA-L

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Dexamethasone is a glucocorticoid and an anti-inflammatory agent, widely used in ocular inflammatory diseases.[1][2]
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Dexamethasone Sodium Phosphate may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using differential pulse polarography technique[1] and high-performance liquid chromatography technique.[2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1690 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trace determination of dexamethasone sodium phosphate in pharmaceutical formulations by differential pulse polarography
Jeyaseelan C and Joshi PA
Analytical and Bioanalytical Chemistry, 373(8), 772-776 (2002)
Simultaneous determination of gatifloxacin and dexamethasone sodium phosphate in bulk and pharmaceutical formulations by HPLC
Sireesha RK, et al.
African Journal of Pharmacy and Pharmacology, 5(17), 1990-1995 (2011)
Tzu-Chieh Chen et al.
The Journal of biological chemistry, 294(23), 9213-9224 (2019-05-06)
Chronic or excess glucocorticoid exposure causes lipid disorders such as hypertriglyceridemia and hepatic steatosis. Angptl4 (angiopoietin-like 4), a primary target gene of the glucocorticoid receptor in hepatocytes and adipocytes, is required for hypertriglyceridemia and hepatic steatosis induced by the synthetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service