Skip to Content
Merck
All Photos(1)

Key Documents

PHR1021

Supelco

Clindamycin Phosphate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Clindamycin 2-phosphate, Clindamycin 2-dihydrogen phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€85.20

€85.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€85.20

About This Item

Empirical Formula (Hill Notation):
C18H34ClN2O8PS
CAS Number:
Molecular Weight:
504.96
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€85.20


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C2269000
traceable to USP 1138008

API family

clindamycin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

CCC[C@@H]1C[C@H](N(C)C1)C(=O)NC([C@H](C)Cl)C2O[C@H](SC)[C@H](OP(O)(O)=O)[C@@H](O)[C@H]2O

InChI

1S/C18H34ClN2O8PS/c1-5-6-10-7-11(21(3)8-10)17(24)20-12(9(2)19)15-13(22)14(23)16(18(28-15)31-4)29-30(25,26)27/h9-16,18,22-23H,5-8H2,1-4H3,(H,20,24)(H2,25,26,27)/t9-,10+,11-,12?,13+,14-,15?,16+,18+/m0/s1

InChI key

UFUVLHLTWXBHGZ-MWBQRTRKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Clindamycin is a common antibiotic drug generally employed for the treatment of infections caused by gram-positive aerobes and both gram-negative and gram-positive anaerobic pathogens.[1]

Application

Clindamycin Phosphate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography[2] and potentiometry[3] techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Spectrum of Activity: Gram positive cocci and taxoplasma. Especially active against anaerobic bacteria.
Mode of Action: Inhibits protein synthesis in bacterial by binding the 50s ribosomal subunit.

Other Notes

Antibacterial and antiprotozoal antibiotic of the licosamide class.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC4009 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Lact. - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatography of clindamycin antibiotics using tris (bipyridine)-ruthenium (III) chemiluminescence detection
Targove MA and Danielson ND
Journal of Chromatographic Science, 28(10), 505-509 (1990)
Nonisothermal stability assessment of stable pharmaceuticals: testing of a clindamycin phosphate formulation
Kipp JE and Hlavaty JJ
Pharmaceutical Research, 8(5), 570-575 (1991)
Sub-second adsorption for the fast sub-nanomolar monitoring of Clindamycin in its pure and pharmaceutical samples by fast Fourier transformation with the use of continuous cyclic voltammetry at an Au microelectrode in a flowing system
Norouzi P, et al.
Materials Science and Engineering, C, 28(1), 87-93 (2008)
Diane Thiboutot et al.
Journal of the American Academy of Dermatology, 59(5), 792-800 (2008-09-23)
We sought to evaluate efficacy, safety, and tolerability of a combination of clindamycin phosphate 1.2% and benzoyl peroxide 2.5% (clindamycin-BPO 2.5%) aqueous gel in moderate to severe acne vulgaris. A total of 2813 patients, aged 12 years or older, were
M Badawy Abdel-Naser et al.
Expert opinion on pharmacotherapy, 9(16), 2931-2937 (2008-10-22)
Clindamycin phosphate 1.2% together with tretinoin 0.025% as a gel (CTG) is a topical formulation of a fixed and stable combination approved by the FDA for the treatment of acne vulgaris in patients 12 years of age or older. The

Questions

  1. Hello, Which factor can be used to calculate Clindamycine base in a product having a reference this standar PHR1021 Clindamycine Phosphate? Can I use the stoichiometric factor 424.98/504.96?

    1 answer
    1. Yes, stoichiometric chemistry will be an appropriate method. The molecular weight can be used to calculate clindamycin in clindamycin phosphate. See the link below to review a sample Certificate of Analysis. Page 2 reports the purity of clindamycin as well as clindamycin phosphate.
      https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/rtc/PHR1021_LRAD2296.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service