Skip to Content
Merck
All Photos(1)

Key Documents

PHR1660

Supelco

Indapamide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Indapamide, N-(4-Chloro-3-sulfamoylbenzamido)-2-methylindoline

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€102.00

€102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€102.00

About This Item

Empirical Formula (Hill Notation):
C16H16ClN3O3S
CAS Number:
Molecular Weight:
365.83
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€102.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 999
traceable to Ph. Eur. I0150000
traceable to USP 1338801

API family

indapamide

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC1Cc2ccccc2N1NC(=O)c3ccc(Cl)c(c3)S(N)(=O)=O

InChI

1S/C16H16ClN3O3S/c1-10-8-11-4-2-3-5-14(11)20(10)19-16(21)12-6-7-13(17)15(9-12)24(18,22)23/h2-7,9-10H,8H2,1H3,(H,19,21)(H2,18,22,23)

InChI key

NDDAHWYSQHTHNT-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Indapamide is a diuretic that is categorized under the family of benzothiadiazines compounds.[1] It is an oral drug that is usually prescribed as an antihypertensive agent for patients with mild to moderate hypertension.[2]

Application

Indapamide may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations[3] by various chromatography techniques[1][4][3] and spectrophotometry.[4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of indapamide in pharmaceuticals and urine by high-performance liquid chromatography with amperometric detection.
Legorburu MJ, et al.
Journal of Chromatographic Science, 37(8), 283-287 (1999)
Determination of certain drugs in binary mixtures formulations by second derivative ratio spectrophotometry and LC.
El-Gindy A, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 59(9), 703-712 (2004)
Development of new spectrophotometric methods for the determination of Indapamide in bulk and Pharmaceutical formulations.
Pai JB, et al.
International Journal of ChemTech Research, 3(2), 755-760 (2011)
Comparison of spectrophotometric and an LC method for the determination perindopril and indapamide in pharmaceutical formulations.
Erk N
Journal of Pharmaceutical and Biomedical Analysis, 26(1), 43-52 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service