Skip to Content
Merck
All Photos(1)

Documents

L0399900

Levocarnitine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Carnitine inner salt, (−)-(R)-3-Hydroxy-4-(trimethylammonio)butyrate, Vitamin BT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2CH(OH)CH2COO-
CAS Number:
Molecular Weight:
161.20
Beilstein:
4292315
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levocarnitine

manufacturer/tradename

EDQM

mp

197-212 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[N+](C)(C)C[C@H](O)CC([O-])=O

InChI

1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/t6-/m1/s1

InChI key

PHIQHXFUZVPYII-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levocarnitine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Carnitine is a quaternary amine that occurs naturally in most mammalian tissue. It is present in relatively high concentrations in skeletal muscle and heart where it is involved in regulating energy metabolism. It shifts glucose metabolism from glycolysis to glycogen storage and enhances the transport of long chain fatty acids into the mitochondria where they are oxidized for energy production.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Andrea Lenzi et al.
Fertility and sterility, 81(6), 1578-1584 (2004-06-15)
To determine the efficacy of combined l-carnitine and l-acetyl-carnitine therapy in infertile males with oligo-astheno-teratozoospermia. Placebo-controlled double-blind randomized trial. University tertiary referral center. Sixty infertile patients (aged 20-40 years) with the following baseline sperm selection criteria: concentration, 10 to 40
N A Oey et al.
Placenta, 27(8), 841-846 (2005-11-23)
Carnitine plays an indispensable role in fatty acid oxidation. Previous studies revealed that fetal carnitine is derived from the mother via transplacental transfer. Recent studies demonstrated the presence and importance of an active fatty acid oxidation system in the human
Allan M Evans et al.
Clinical pharmacokinetics, 42(11), 941-967 (2003-08-12)
L-Carnitine is a naturally occurring compound that facilitates the transport of fatty acids into mitochondria for beta-oxidation. Exogenous L-carnitine is used clinically for the treatment of carnitine deficiency disorders and a range of other conditions. In humans, the endogenous carnitine
Xiaoshan Zhou et al.
PloS one, 8(3), e58843-e58843 (2013-03-19)
Thymidine kinase 2 (TK2) deficiency in humans causes mitochondrial DNA (mtDNA) depletion syndrome. To study the molecular mechanisms underlying the disease and search for treatment options, we previously generated and described a TK2 deficient mouse strain (TK2(-/-)) that progressively loses
Sandra Wächter et al.
Clinica chimica acta; international journal of clinical chemistry, 318(1-2), 51-61 (2002-03-07)
Long-term administration of high oral doses of L-carnitine on the skeletal muscle composition and the physical performance has not been studied in humans. Eight healthy male adults were treated with 2 x 2 g of L-carnitine per day for 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service