Skip to Content
Merck
All Photos(1)

Key Documents

94954

Supelco

L-Carnitine hydrochloride

analytical standard

Synonym(s):

(−)-β-Hydroxy-γ-(trimethylammonio)butyrate, Vitamin BT

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€109.00

€109.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€109.00

About This Item

Linear Formula:
(CH3)3N+CH2CH(OH)CH2CO2H · Cl-
CAS Number:
Molecular Weight:
197.66
MDL number:
UNSPSC Code:
41116107
NACRES:
NB.21

€109.00


Estimated to ship on26 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]/D -24.0±2.0°, c = 0.2 in H2O

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O

InChI

1S/C7H15NO3.ClH/c1-8(2,3)5-6(9)4-7(10)11;/h6,9H,4-5H2,1-3H3;1H/t6-;/m1./s1

InChI key

JXXCENBLGFBQJM-FYZOBXCZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Carnitine is a quaternary amine that occurs naturally in most mammalian tissue. It is present in relatively high concentrations in skeletal muscle and heart where it is involved in regulating energy metabolism. It shifts glucose metabolism from glycolysis to glycogen storage and enhances the transport of long chain fatty acids into the mitochondria where they are oxidized for energy production.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service