Skip to Content
Merck
All Photos(1)

Key Documents

F0060000

Fenticonazole nitrate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21Cl2N3O4S
CAS Number:
Molecular Weight:
518.41
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

fenticonazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

S(c2ccc(cc2)COC(C[n]4cncc4)c3c(cc(cc3)Cl)Cl)c1ccccc1.[N+](=O)([O-])O

InChI

1S/C24H20Cl2N2OS.HNO3/c25-19-8-11-22(23(26)14-19)24(15-28-13-12-27-17-28)29-16-18-6-9-21(10-7-18)30-20-4-2-1-3-5-20;2-1(3)4/h1-14,17,24H,15-16H2;(H,2,3,4)

InChI key

FJNRUWDGCVDXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fenticonazole nitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srpski arhiv za celokupno lekarstvo, 140(7-8), 469-474 (2012-10-25)
Uncomplicated vulvovaginal candidiasis appears in 75% women of reproductive age. The most frequent causes are Candida albicans (85-95%) or C. glabrata, and infrequently C. krusei, C. tropicalis, C. parapsilosis, C. pseudotropicalis, etc. The aim of the study was to investigate
F Murina et al.
Minerva ginecologica, 64(2), 89-94 (2012-04-07)
The aim of this paper was to compare the efficacy of fluconazole 150 mg and intra-vaginal fenticonazole 600mg in short-course treatment of the acute episode of vulvovaginal candidiasis (VVC). In a prospective study, 80 patients with clinical and mycological (SavvyCheck™
P Altmeyer et al.
The Journal of international medical research, 18(1), 61-67 (1990-01-01)
A double-blind clinical trial was performed to evaluate efficacy and tolerance of once-daily 2% fenticonazole compared with 1% cyclopyroxolamine spray applied for 2-4 weeks in 100 patients with cutaneous mycotic lesions. After treatment lasting 21.9 +/- 6.7 or 22.5 +/-
A M Di Pietra et al.
Journal of pharmaceutical and biomedical analysis, 10(10-12), 873-879 (1992-10-01)
Reversed-phase HPLC on different column packing materials (Hypersil C-18, Spherisorb-CN, Chromspher-B) is used to obtain selective separations of imidazole antimycotic drugs, such as ketoconazole, clotrimazole, tioconazole, bifonazole, isoconazole, econazole, miconazole and fenticonazole. The use of a post-column on-line photochemical reactor
A G Lawrence et al.
Current medical research and opinion, 12(2), 114-120 (1990-01-01)
An open, randomized comparative clinical trial was performed in 153 patients suffering from symptomatic vaginal candidiasis confirmed by mycological tests. Patients were allocated at random into two groups: the first group (consisting of 75 subjects) was treated with a single

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service