Skip to Content
Merck
All Photos(2)

Key Documents

1.09990

Supelco

Silver nitrate solution

c(AgNO₃) = 0.1 mol/l (0.1 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
€99.90

€99.90


Estimated to ship on29 May 2025



Select a Size

Change View
1 AMP
€99.90

About This Item

Empirical Formula (Hill Notation):
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

€99.90


Estimated to ship on29 May 2025


Product Name

Silver nitrate solution, for 1000 ml, c(AgNO₃) = 0.1 mol/l (0.1 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Precipitation reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

7-9 (20 °C in H2O)

density

1.27 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[Ag]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Antibacterial polymeric vesicles: A novel method for the in situ fabrication of silver nanoparticle-decorated polymeric vesicles was developed using silver nitrate, indicating significant antibacterial properties suitable for various biomedical applications (Zhang et al., 2024).

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service