Skip to Content
Merck

Skip To

CL10

CCS Viscosity Standard

ASTM D 5293 and SAE Specification J304

Synonym(s):

Cold-Cranking Simulator Viscosity Standard

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107


grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon CCS Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.


Still not finding the right product?

Explore all of our products under CCS Viscosity Standard


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Paula Grazielle Chaves da Silva et al.
Cellular and molecular neurobiology, 40(6), 967-989 (2020-01-26)
Decapod crustaceans, like mammals, retain the ability to make new neurons throughout life. In mammals, immune cells are closely associated with stem cells that generate adult-born neurons. In crayfish, evidence suggests that immune cells (hemocytes) originating in the immune system
Arun Mouli Kolinjivadi et al.
Molecular cell, 67(5), 867-881 (2017-08-02)
Brca2 deficiency causes Mre11-dependent degradation of nascent DNA at stalled forks, leading to cell lethality. To understand the molecular mechanisms underlying this process, we isolated Xenopus laevis Brca2. We demonstrated that Brca2 protein prevents single-stranded DNA gap accumulation at replication
Amaia Ercilla et al.
Cell reports, 30(7), 2416-2429 (2020-02-23)
It has been long assumed that normally leading strand synthesis must proceed coordinated with the lagging strand to prevent strand uncoupling and the pathological accumulation of single-stranded DNA (ssDNA) in the cell, a dogma recently challenged by in vitro studies in



Global Trade Item Number

SKUGTIN
CL10-500ML04061837679964

Questions

Reviews

No rating value

Active Filters