Skip to Content
Merck
All Photos(1)

Documents

B0700000

Benzylpenicillin potassium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Penicillin G potassium salt, Benzylpenicillin potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17KN2O4S
CAS Number:
Molecular Weight:
372.48
Beilstein:
3832841
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

penicillin & derivatives

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[K+].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)Cc3ccccc3)C([O-])=O

InChI

1S/C16H18N2O4S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1

InChI key

IYNDLOXRXUOGIU-LQDWTQKMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzylpenicillin potassium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mode of Action: Penicillin G acts by inhibiting cell wall synthesis through binding to penicillin binding proteins (PBPs), inhibiting peptidoglycan chain cross-linking.

Antimicrobial spectrum: This product is active against gram-positive and gram-negative bacteria.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthieu Picard et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 113(1), 75-81 (2014-05-27)
The absence of commercially available penicilloyl-polylysine (PPL) for most of the last decade severely hampered the practice of penicillin allergy evaluation because skin testing without PPL is reported to have a poor negative predictive value (NPV). To determine the safety
Nahid Pourreza et al.
Journal of AOAC International, 97(4), 1225-1229 (2014-08-26)
A cloud point extraction (CPE) method for determination of trace amounts of penicillin G by spectrophotometry based on its effect on the triiodide ion (I3(-)) has been developed. Penicillin G is converted to the corresponding penicilloic acid by carrying out
Chronic inflammatory demyelinating polyneuropathy associated with neurosyphilis.
Evangelia Kararizou et al.
The American journal of the medical sciences, 349(1), 90-91 (2014-12-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service