Skip to Content
Merck
All Photos(1)

Key Documents

95710

Sigma-Aldrich

Sodium xylenesulfonate

technical, mixture of isomers, ≥90% (T)

Synonym(s):

Dimethylbenzenesulfonic acid sodium salt, Xylenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€128.00

€128.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€128.00

About This Item

Linear Formula:
(CH3)2C6H3SO3Na
CAS Number:
Molecular Weight:
208.21
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

€128.00


Check Cart for Availability

Request a Bulk Order

Quality Level

grade

technical

Assay

≥90% (T)

form

powder

SMILES string

[Na+].Cc1ccc(cc1C)S([O-])(=O)=O

InChI

1S/C8H10O3S.Na/c1-6-3-4-8(5-7(6)2)12(9,10)11;/h3-5H,1-2H3,(H,9,10,11);/q;+1/p-1

InChI key

QUCDWLYKDRVKMI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyan Mou et al.
Bioresource technology, 220, 637-640 (2016-09-04)
The objective of this study was to investigate the effects of organosolv and hydrotropic pretreatments on improving enzymatic hydrolysis of eucalyptus. The chemical composition of the fiber surface was analyzed using X-ray photoelectron spectroscopy (XPS) to determine the surface characteristics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service