Skip to Content
Merck

Skip To

242535

Sodium xylenesulfonate solution

mixture of isomers, 40 wt. % in H2O

Synonym(s):

Sodium dimethylbenzenesulfonate, Sodium xylenesulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2C6H3SO3Na
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

concentration

40 wt. % in H2O

refractive index

n20/D 1.405

density

1.17 g/mL at 25 °C

functional group

sulfonic acid

SMILES string

[Na+].[S](=O)(=O)([O-])c1cc(c(cc1)C)C

InChI

1S/C8H10O3S.Na/c1-6-3-4-8(5-7(6)2)12(9,10)11;/h3-5H,1-2H3,(H,9,10,11);/q;+1/p-1

InChI key

QUCDWLYKDRVKMI-UHFFFAOYSA-M

Application

Sodium xylenesulfonate solution may be used in chemical synthesis studies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
3357387151379724
description

mixture of isomers, 40 wt. % in H2O

description

1.0 M in THF

description

~40% in H2O

description

1 M, suitable for HPCE

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

refractive index

n20/D 1.405

refractive index

-

refractive index

-

refractive index

-

density

1.17 g/mL at 25 °C

density

0.897 g/mL at 25 °C

density

-

density

-

form

liquid

form

liquid

form

liquid

form

liquid

concentration

40 wt. % in H2O

concentration

1.0 M in THF

concentration

~40% in H2O

concentration

0.9-1.1 M

functional group

sulfonic acid

functional group

-

functional group

amine, dithiocarbamate

functional group

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters