Skip to Content
Merck
All Photos(1)

Key Documents

71220

Sigma-Aldrich

Sodium 2-ethylhexyl sulfate

~50% in H2O

Synonym(s):

2-Ethylhexyl sulfate sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€99.80

€99.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€99.80

About This Item

Empirical Formula (Hill Notation):
C8H17NaO4S
CAS Number:
Molecular Weight:
232.27
Beilstein:
5177087
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€99.80


Check Cart for Availability

Request a Bulk Order

description

anionic

form

liquid

mol wt

232.27 g/mol

concentration

4.95% (Na, ICP)
~50% in H2O

technique(s)

protein quantification: suitable

density

1.12 g/mL at 20 °C (lit.)

SMILES string

[Na+].CCCCC(CC)COS([O-])(=O)=O

InChI

1S/C8H18O4S.Na/c1-3-5-6-8(4-2)7-12-13(9,10)11;/h8H,3-7H2,1-2H3,(H,9,10,11);/q;+1/p-1

InChI key

DGSDBJMBHCQYGN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 2-ethylhexyl sulfate is an anionic surfactant that can be used:
  • In suspension polymerization.[1]
  • In the analysis of phenolic compounds through microchip-CE with pulsed amperometric detection.[2]
  • As charge balancing anions in the synthesis of organo-layered double hydroxides (organo-LDHs).[3]

Other Notes

Alkyl sodium sulfate surfactants.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of alkyl sulfates on the analysis of phenolic compounds by microchip capillary electrophoresis with pulsed amperometric detection.
Ding Y, et al.
Analyst, 132(10), 997-1004 (2007)
Variation of anions in layered double hydroxides: effects on dispersion and fire properties.
Wang L, et al.
Polymer Degradation and Stability, 94(5), 770-781 (2009)
Suspension polymerization of thermally expandable core/shell particles.
Jonsson M, et al.
Polymer, 47(10), 3315-3324 (2006)
W M Kluwe
Environmental health perspectives, 65, 271-278 (1986-03-01)
Chronic toxicity and carcinogenicity studies of several phthalic acid esters (PAEs) and compounds containing a 2-ethylhexyl moiety were conducted in Fischer 344 rats and B6C3F1 (hybrid) mice. The compounds studied were phthalic anhydride, di(2-ethylhexyl) phthalate, butyl benzyl phthalate, diallyl phthalate

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service