Skip to Content
Merck
All Photos(1)

Key Documents

93303

Sigma-Aldrich

Tris(2-aminoethyl)amine

technical, ≥90% (GC)

Synonym(s):

2,2′,2′′-Nitrilotriethylamine, 2,2′,2′′-Triaminotriethylamine, TAEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2CH2CH2)3N
CAS Number:
Molecular Weight:
146.23
Beilstein:
1739626
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

5 (vs air)

vapor pressure

0.02 mmHg ( 20 °C)

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.497 (lit.)
n20/D 1.497

bp

114 °C/15 mmHg (lit.)

density

0.976 g/mL at 20 °C (lit.)

SMILES string

NCCN(CCN)CCN

InChI

1S/C6H18N4/c7-1-4-10(5-2-8)6-3-9/h1-9H2

InChI key

MBYLVOKEDDQJDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Scarso et al.
The Journal of organic chemistry, 72(2), 376-385 (2007-01-16)
Three artificial amino acids derived from l-serine by replacing the hydroxyl moiety with 1,4,7-triazacyclononane, 1,5,9-triazacyclododecane, and 1,4,7,10-tetraazacyclododecane, respectively, have been connected to the three arms of the tetraamine tris(2-aminoethyl)amine, Tren, to obtain tripodal ligands. They are able to bind up
Paloma Arranz et al.
Inorganic chemistry, 49(20), 9321-9332 (2010-09-15)
Binding of anions of great environmental concern such as SO(4)(2-), PO(4)(3-), AsO(4)(3-), HgCl(4)(2-), and CrO(4)(2-) by the protonated forms of a tren-like (tren = tris(2-aminoethyl)amine) ligand (HL) functionalized with a pyrimidine residue was studied by means of potentiometric measurements and
Pascal Dimitrov Raytchev et al.
The Journal of organic chemistry, 75(6), 2099-2102 (2010-02-20)
The first hemicryptophanes derived from tris(N-alkyl-carbamoylmethyl)amine and tris(2-aminoethyl)amine (tren) have been synthesized following a single synthetic pathway that allows the subsequent formation of the two heteroditopic hosts 3 and 4. X-ray crystal structures show a well-defined cavity encapsulating a solvent
Cynthia T Brewer et al.
Dalton transactions (Cambridge, England : 2003), 40(1), 181-194 (2010-11-26)
Reaction of H(3)L(1), the Schiff base condensate of tris(2-aminoethyl)amine with three equivalents of 5-methyl-1H-pyrazole-3-carboxaldehyde, with manganese(II)perchlorate or iron(II)tetrafluoroborate results in the isolation of [MH(3)L(1)]X(2) (M = Mn and X = ClO(4) and M = Fe and X = BF(4)). These
Sulfate recognition by persistent crystalline capsules with rigidified hydrogen-bonding cavities.
Radu Custelcean et al.
Angewandte Chemie (International ed. in English), 47(10), 1866-1870 (2008-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service