Skip to Content
Merck
All Photos(1)

Key Documents

92412

Millipore

Yeast Mannitol Agar without Calcium

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€187.00

€187.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€187.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

€187.00


Estimated to ship on14 May 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
dipotassium phosphate, 0.5 g/L
magnesium sulfate, 0.2 g/L
mannitol, 10 g/L
sodium chloride, 0.1 g/L
yeast extract, 1 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

6.8±0.2 (25 °C)

application(s)

agriculture
life science and biopharma

microbiology

suitability

rhizobiaceae

Application

Used for cultivation, isolation and enumeration of soil microorganisms like Rhizobium.

Preparation Note

Suspend 26.8 g of Yeast Mannitol Agar without Calcium in 1000 ml of distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service