Skip to Content
Merck
All Photos(2)

Key Documents

1.05982

Millipore

D-Mannitol

suitable for microbiology

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€92.30

€92.30


Estimated to ship on26 May 2025


Request more information

Select a Size

Change View
500 G
€92.30

About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
MDL number:
UNSPSC Code:
41106212
EC Index Number:
200-711-8
NACRES:
NA.74

€92.30


Estimated to ship on26 May 2025


Request more information

autoignition temp.

410 °C

Quality Level

potency

13500 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 100 g/L in H2O)

bp

290-295 °C/4 hPa

mp

167-170 °C (lit.)

solubility

213 g/L

bulk density

400‑500 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Analysis Note

Identity (IR-spectrum): passes test
Chloride (Cl): ≤ 0.003 %
Sulfate (SO₄): ≤ 0.01
Water: ≤ 0.5 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service