Skip to Content
Merck
All Photos(1)

Key Documents

91299

Supelco

Isobutyryl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-(2-methyl-1-oxopropoxy)-1-propanaminium inner salt, L-Carnitine isobutyryl ester, iC4-Carnitine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€124.00

€124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€124.00

About This Item

Empirical Formula (Hill Notation):
C11H21NO4
CAS Number:
Molecular Weight:
231.29
Beilstein:
4138517
MDL number:
UNSPSC Code:
41116107
NACRES:
NB.21

€124.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]/D -22.0±2.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤10.0% water (calculated from elemental analysis)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(C(C)C)=O)CC([O-])=O

InChI

1S/C11H21NO4/c1-8(2)11(15)16-9(6-10(13)14)7-12(3,4)5/h8-9H,6-7H2,1-5H3/t9-/m1/s1

InChI key

LRCNOZRCYBNMEP-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobutyryl-L-carnitine belongs to the acylcarnitines class of compounds.[1] These compounds are produced from L-carnitine, which plays an important role in fatty acid metabolic pathways. The functions and roles of acyl-L-carnitines in various tissues like brain, heart, and muscle continue to attract much interest.

Application

Isobutyryl-L-carnitine may be used as an analytical reference standard for the separation and identification of underivatized isobutyryl-L-carnitine in human plasma samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS).[1] It may also be used as an analytical reference standard for the quantification of the analyte in human urine samples using liquid chromatography/electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS).[2]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix effect?corrected liquid chromatography/tandem mass-spectrometric method for determining acylcarnitines in human urine
Abe K, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 468, 187-194 (2017)
Minzhi Peng et al.
Journal of chromatography. A, 1319, 97-106 (2013-10-31)
A simple HPLC-MS/MS method has been established to separate and identify underivatized acylcarnitine isomers. Human plasma samples were deproteinized and concentrated. Acylcarnitines were separated on a reverse phase column and detected with triple quadrupole linear ion trap mass spectrometry. Deuterium-labeled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service