Skip to Content
Merck

Skip To

08984

Butyryl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxobutoxy)-1-propanaminium inner salt, Butanoyl-L-carnitine, C4-Carnitine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 121,00

About This Item

Empirical Formula (Hill Notation):
C11H21NO4
CAS Number:
Molecular Weight:
231.29
UNSPSC Code:
41116107
NACRES:
NB.21
MDL number:

€ 121,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (HPLC)

optical activity

[α]/D -23.0±2.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤10.0% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCC)=O)CC([O-])=O

InChI

1S/C11H21NO4/c1-5-6-11(15)16-9(7-10(13)14)8-12(2,3)4/h9H,5-8H2,1-4H3/t9-/m1/s1

InChI key

QWYFHHGCZUCMBN-SECBINFHSA-N

General description

Butyryl-L-carnitine is a naturally occurring compound belonging to the acylcarnitine class of compounds as listed in the human metabolome database (HMBD). These compounds are reported to be used as biomarkers in the diagnosis of carnitine deficiency. It acts as a carrier of long chain acyl groups from activated fatty acids across the inner mitochondrial membrane into the mitochondrial matrix where they undergo β-oxidation to acetyl CoA to obtain usable energy via the citric acid cycle.

Application

Butyryl-L-carnitine may be used as an analytical reference standard for the separation and identification of underivatized butyryl-L-carnitine in human plasma samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS). It may also be used as an analytical reference standard for the quantification of the analyte in biological samples[1] using chromatography-based techniques.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
080844262391299
application(s)

clinical testing

application(s)

clinical testing

application(s)

-

application(s)

clinical testing

grade

analytical standard

grade

analytical standard

grade

-

grade

analytical standard

assay

≥97.0% (HPLC)

assay

≥97.0% (HPLC)

assay

≥97.0% (TLC)

assay

≥97.0% (HPLC)

format

neat

format

neat

format

-

format

neat

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

impurities

≤10.0% water

impurities

≤10.0% water

impurities

-

impurities

≤10.0% water (calculated from elemental analysis)


Still not finding the right product?

Explore all of our products under Butyryl-L-carnitine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters