Skip to Content
Merck

Skip To

91019

Schaedler Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Schaedler Anaerobe Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 g
Check Cart for Availability
€ 199,00

About This Item

NACRES:
NA.85
UNSPSC Code:
41171606

€ 199,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L , casein enzymic hydrolysate, 5.67 g/L , L-cystine, 0.40 g/L , dextrose, 5.83 g/L , dipotassium hydrogen phosphate, 0.83 g/L , hemin, 0.010 g/L , papaic digest of soyabean meal, 1.0 g/L , proteose peptone, 5.0 g/L , sodium chloride, 1.67 g/L , tris hydroxymethyl aminomethane, 3.0 g/L , yeast extract, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.6±0.2 (25 °C)

application(s)

clinical testing
food and beverages, microbiology

suitability

anaerobic bacteria, nonselective for bacteria (General Media)

Application

Schaedler Agar is used for the enumeration of various aerobic and anaerobic bacterial species present in the gastrointestinal tract.

Preparation Note

Suspend 43.41 grams in 950 ml distilled water. Heat to boiling with frequent agitation to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool and add 5% sterile defibrinated blood if desired. Mix well before dispensing. Avoid overheating and photo oxidation of the medium, as it will retard the growth of bacteria.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
843688564052062
suitability

anaerobic bacteria, nonselective for bacteria (General Media)

suitability

selective and differential for Citrobacter spp., selective and differential for Escherichia coli, selective and differential for Klebsiella spp., selective and differential for Proteus spp., selective and differential for Salmonella spp., selective and differential for Shigella spp., selective and differential for coliforms, selective and differential for enterobacteriaceae

suitability

selective for Salmonella spp., selective for Shigella spp

suitability

nonselective for Escherichia coli, nonselective for bacteria (General Media), nonselective for coliforms

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

form

powder

form

powder

form

powder

form

powder

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

composition

agar, 15.0 g/L , L-cystine, 0.40 g/L , dipotassium hydrogen phosphate, 0.83 g/L , papaic digest of soyabean meal, 1.0 g/L , sodium chloride, 1.67 g/L , yeast extract, 5.0 g/L , casein enzymic hydrolysate, 5.67 g/L , hemin, 0.010 g/L , tris hydroxymethyl aminomethane, 3.0 g/L , dextrose, 5.83 g/L , proteose peptone, 5.0 g/L

composition

agar, 15.0 g/L , bile salt mixture, 3.825 g/L , brilliant green, 0.0006 g/L , disodium hydrogen phosphate 2 hydrate, 1.0 g/L , ferric citrate, 0.5 g/L , lactose, 11.5 g/L , magnesium sulfate, 0.4 g/L , meat extract, 6.0 g/L , meat peptone, 3.8 g/L , metachrome yellow, 0.47 g/L , methyl blue, 0.25 g/L , neutral red, 0.022 g/L , L-phenylalanine, 5.0 g/L , sodium thiosulfate 5 hydrate, 4.25 g/L , starch (soluble), 0.5 g/L , sucrose, 13.0 g/L , trisodium citrate5.5 hydrate, 9.3 g/L , yeast extract, 3.0 g/L

composition

agar, 15.0 g/L , brilliant green, 0.0003 g/L , ferric citrate, 1.0 g/L , lactose, 10.0 g/L , meat extract, 5.0 g/L , neutral red, 0.025 g/L , ox bile, dehydrated, 8.5 g/L , peptone, 5.0 g/L , sodium citrate, 10.0 g/L , sodium thiosulfate, 8.5 g/L

composition

agar, 15 g/L , casein enzymic hydrolysate, 10 g/L , sodium chloride, 10 g/L , yeast extract, 5 g/L

application(s)

clinical testing
food and beverages, microbiology

application(s)

clinical testing
environmental
food and beverages, microbiology

application(s)

clinical testing
environmental
food and beverages, microbiology

application(s)

clinical testing
food and beverages
life science and biopharma, microbiology


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
SML1137-25MG04061832983066
SML1137-5MG04061832983073
530583000104055977260915
91019-500G04061833254691

Questions

Reviews

No rating value

Active Filters