Skip to Content
Merck
All Photos(2)

Key Documents

901813

Sigma-Aldrich

SnatchCat

≥95%

Synonym(s):

1,4-Bis(3-isocyanopropyl)piperazine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€119.00

€119.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€119.00

About This Item

Empirical Formula (Hill Notation):
C12H20N4
CAS Number:
Molecular Weight:
220.31
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€119.00


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: catalyst
reagent type: chelator

mp

82-83 °C

storage temp.

2-8°C

SMILES string

[N+](#[C-])CCCN1CCN(CC1)CCC[N+]#[C-]

InChI key

GTYUVOWMIHCPRM-UHFFFAOYSA-N

Application

SnatchCat is a universal metal scavenger with a proven efficiency for Ru and Pd metals. The scavenger is a metathesis reaction quencher, reducing metal content to 10 ppm after 20 min of scavenging time followed by silica gel filtration. SnatchCat is compatible with a broad range of functional groups and solvents. The product is stable, non-volatile and odour-free.

Legal Information

Sold in collaboration with Apeiron Synthesis, Inc. Research quantities are 100 g / customer / year. For quantities that exceed the research limit, please contact the supplier at www.apeiron-synthesis.com.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grzegorz Szczepaniak et al.
ChemSusChem, 8(24), 4139-4148 (2015-11-12)
Three isocyanides containing a tertiary nitrogen atom were investigated for use as small-molecule ruthenium scavenging agents in the workup of olefin metathesis reactions. The proposed compounds are odorless, easy to obtain, and highly effective in removing metal residues, sometimes bringing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service