Skip to Content
Merck
All Photos(3)

Key Documents

577944

Sigma-Aldrich

Hoveyda-Grubbs Catalyst® M700

Umicore

Synonym(s):

Hoveyda-Grubbs Catalyst® 1st Generation, Hoveyda-Grubbs Catalyst® M70 (C601), Dichloro(2-isopropoxyphenylmethylene) (tricyclohexylphosphine)ruthenium(II), Dichloro(o-isopropoxyphenylmethylene)(tricyclohexylphosphine)ruthenium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€28.90
500 MG
€82.10
2 G
€241.00
5 G
€532.00

€28.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€28.90
500 MG
€82.10
2 G
€241.00
5 G
€532.00

About This Item

Empirical Formula (Hill Notation):
C28H45Cl2OPRu
CAS Number:
Molecular Weight:
600.61
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€28.90


Check Cart for Availability

Request a Bulk Order

Quality Level

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Ring Opening Metathesis Polymerisation

mp

195-197 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]\C(c1ccccc1OC(C)C)=[Ru](\Cl)Cl.C2CCC(CC2)P(C3CCCCC3)C4CCCCC4

InChI

1S/C18H33P.C10H12O.2ClH.Ru/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-8(2)11-10-7-5-4-6-9(10)3;;;/h16-18H,1-15H2;3-8H,1-2H3;2*1H;/q;;;;+2/p-2

InChI key

KMKCJXPECJFQPQ-UHFFFAOYSA-L

Application

Proven to be useful in Ring-Closing Metathesis (RCM) to form macrocycles. Also employed in a pivotal, macrocyclic ring-closing metathesis in the multi-step synthesis of an HCV protease inhibitor.

Learn more about our metathesis catalysts

Legal Information

Product of Umicore

Additional information available at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan K Yee et al.
The Journal of organic chemistry, 71(19), 7133-7145 (2006-09-09)
A multistep scalable synthesis of the clinically important hepatitis C virus (HCV) protease inhibitor BILN 2061 (1) is described. The synthesis is highly convergent and consists of two amide bond formations, one etherification, and one ring-closing metathesis (RCM) step, using
Sequential catalysis: a metathesis/dihydroxylation sequence.
Samuel Beligny et al.
Angewandte Chemie (International ed. in English), 45(12), 1900-1903 (2006-02-14)
Schrodi, Y.; Pederson, R. L.
Aldrichimica Acta, 45-45 (2007)
T M Trnka et al.
Accounts of chemical research, 34(1), 18-29 (2001-02-15)
In recent years, the olefin metathesis reaction has attracted widespread attention as a versatile carbon-carbon bond-forming method. Many new applications have become possible because of major advances in catalyst design. State-of-the-art ruthenium catalysts are not only highly active but also
Ruthenium-based heterocyclic carbene-coordinated olefin metathesis catalysts.
Georgios C Vougioukalakis et al.
Chemical reviews, 110(3), 1746-1787 (2009-12-17)

Related Content

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service