Skip to Content
Merck
All Photos(1)

Key Documents

900673

Sigma-Aldrich

Mixture of metal scavengers on silica gel

≥99%, molecular loading ≥0.45 mmol/g

Synonym(s):

Si-Spectrum, SiliaMets® Spectrum

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€93.00

€93.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€93.00

About This Item

UNSPSC Code:
12161600
NACRES:
NA.22

€93.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

powder

shelf life

2 yr (expiration date period)

reaction suitability

reagent type: catalyst
reagent type: chelator

surface area

480-550 m2/g

pore size

60 Å pore diameter

capacity

≥0.45 mmol/g loading(molecular loading)

Application

It is a proprietary mixture of metal scavengers grafted on silica gel effective in complexing the following: Ca, Cd, Co, Cr, Cs, Cu, Fe, Ir, La, Li, Mg, Ni, Os, Pd, Pt, Rh, Ru, Sc, and Zn.

Legal Information

SiliaMetS is a registered trademark of Silicycle, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service