Skip to Content
Merck
All Photos(1)

Key Documents

76040

Supelco

Palladium Matrix Modifier

Synonym(s):

Palladium(II) nitrate – nitric acid solution, Palladium modifier

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€239.00

€239.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€239.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.21

€239.00


Estimated to ship on24 May 2025


Request a Bulk Order

form

liquid

Quality Level

mol wt

Mr 230.41 g/mol

reaction suitability

reagent type: catalyst
core: palladium

concentration

10.0 g/L±0.3 g/L Pd

cation traces

Ag: ≤0.1 mg/kg
As: ≤0.01 mg/kg
Cd: ≤0.001 mg/kg
Pb: ≤0.01 mg/kg
Se: ≤0.1 mg/kg
Tl: ≤0.02 mg/kg

SMILES string

[Pd].O[N+]([O-])=O

InChI

1S/HNO3.Pd/c2-1(3)4;/h(H,2,3,4);

InChI key

UVCRDXRRYBAMHB-UHFFFAOYSA-N

General description

Matrix modifier in general, is a buffer that is similar to the chemical environment of aqueous standards and samples. It can also make analyte more thermally stable salts, oxides or metal compounds.[1] It also reduces the volatility of analyte.

Application

Palladium matrix modifier may be used in determination of total arsenic by in situ oxidation of diluted urine using graphite furnace atomic absorption spectrophotometer.[2]

Components

Pd(NO3)2.2H2O in 13-20% nitric acid

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauri H. J. Lajunen, P. Peramaki
Spectrochemical Analysis by Atomic Absorption and Emission, 143 -143 (2004)
D E Nixon et al.
Clinical chemistry, 37(9), 1575-1579 (1991-09-01)
We evaluated the effectiveness of nickel and palladium with or without added potassium persulfate as matrix modifiers for the determination of total arsenic in urine. Complete recovery of pure aqueous solutions of As(III), As(V), cacodylic acid (DMA), monomethylarsinic acid (MMA)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service