Skip to Content
Merck
All Photos(2)

Key Documents

73235

Supelco

Sennoside C

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€961.00

€961.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€961.00

About This Item

Empirical Formula (Hill Notation):
C42H40O19
CAS Number:
Molecular Weight:
848.76
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

€961.00


Estimated to ship on24 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2cccc3[C@@H]([C@@H]4c5cccc(O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c5C(=O)c7c(O)cc(cc47)C(O)=O)c8cc(CO)cc(O)c8C(=O)c23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C42H40O19/c43-11-14-7-18-26(16-3-1-5-22(30(16)34(50)28(18)20(46)8-14)58-41-38(54)36(52)32(48)24(12-44)60-41)27-17-4-2-6-23(59-42-39(55)37(53)33(49)25(13-45)61-42)31(17)35(51)29-19(27)9-15(40(56)57)10-21(29)47/h1-10,24-27,32-33,36-39,41-49,52-55H,11-13H2,(H,56,57)/t24-,25-,26?,27?,32-,33-,36+,37+,38-,39-,41-,42-/m1/s1

InChI key

ZFWOUNNKSHIAFK-JZOSXXJUSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sennoside C is an anthraquinone glycoside, belonging to the group of sennosides, widely found in Senna plant. It is commonly used worldwide as a natural laxative.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sennoside C may be used as an analytical reference standard for the quantification of the analyte in Senna and formulations using high-performance liquid chromatography.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

651.9 °F

Flash Point(C)

344.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical studies on the active constituents in crude drugs. IV. Determination of sennosides in senna and formulations by high-performance liquid chromatography
Hayashi I-S, et al.
Chemical & Pharmaceutical Bulletin, 28(2), 120-128 (1980)
Next generation sequencing and transcriptome analysis predicts biosynthetic pathway of sennosides from Senna (Cassia angustifolia Vahl.), a non-model plant with potent laxative properties
Reddy RRN, et al.
Testing, 10(6), 120-128 (2015)
K Yamauchi et al.
The Journal of pharmacy and pharmacology, 44(12), 973-976 (1992-12-01)
Sennosides A and C directly injected into the caecum of mice showed equal purgative activity. Intracaecal administration reduced time to onset of diarrhoea induced by sennoside C from about 3 h after oral administration to about 24 min. At 2.3
T Yagi et al.
The Journal of pharmacy and pharmacology, 49(1), 22-25 (1997-01-01)
This study aimed to explore the mechanism involved in the synergistic purgative action of aloe-emodin anthrone and rhein anthrone, the active metabolites of sennoside C. Aloe-emodin anthrone and rhein anthrone, and their equimolar mixture, induced excretion of an approximately equal
Potentiating effect of sennoside C on purgative activity of sennoside A in mice.
K Kisa et al.
Planta medica, 42(3), 302-303 (1981-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service