Skip to Content
Merck
All Photos(1)

Key Documents

1.07289

Supelco

Palladium matrix modifier

for graphite furnace AAS c(Pd) = 10.0 ± 0.2 g/l (Pd(NO₃)₂ / HNO₃ ca. 15%), for inorganic trace analysis

Synonym(s):

Palladium Interference Reduction Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.21
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

technique(s)

AAS: suitable

density

1.11 g/cm3 at 20 °C

application(s)

general analytical

storage temp.

2-30°C

Application


  • Lead Determination in Complex Matrices: Palladium nanoparticles are employed as chemical modifiers in high-resolution continuum source atomic absorption spectrometry (HR-CS AAS) for determining lead in complex matrices. Recent studies have demonstrated their effectiveness in analyzing lead content in leached extracts from petroleum waste. This application enhances the sensitivity and accuracy of lead detection, crucial for environmental monitoring and industrial waste management (Coêlho et al., International Journal of Environmental Analytical Chemistry, 2022).

Analysis Note

Assay (palladium): 9.8 - 10.2 g/l
Ag (Silver): ≤ 0.000020 g/l
As (Arsenic): ≤ 0.000010 g/l
Cd (Cadmium): ≤ 0.000001 g/l
Pb (Lead): ≤ 0.000010 g/l
Se (Selenium): ≤ 0.00010 g/l
Tl (Thallium): ≤ 0.000020 g/l

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service