Skip to Content
Merck
All Photos(1)

Key Documents

72784

Supelco

Sodium carbonate concentrate

Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7

Synonym(s):

Sodium carbonate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NB.21
Pricing and availability is not currently available.

Quality Level

form

liquid

concentration

72 mM Na2CO3 in H2O

technique(s)

ion chromatography: suitable

SMILES string

[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This eluent concentrate for ion chromatography is determined by potentiometric titration. Content and expiry date can be found on the certificate.

Application

Metrohm IC application note AN-S-345 :ß-Glycerophosphateand L-malate in a pharmaceutical formulation.

Metrohm IC application note AN-S-312 :Disinfectionbyproducts and standard anions according to EPA 300.1 A & B on 930 CompactIC Flex.

Metrohm IC application note AN-S-303 :Anions in KOH (50%) applying Inline Neutralization and intelligent Partial LoopInjection Technique (MiPT).

Linkage

Visit the IC Portal to learn more

Preparation Note

Prepared with sodium carbonate and high purity water (18.2 MΩ, 0.2 μm filtered)

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukiyo Sato et al.
PloS one, 9(6), e99041-e99041 (2014-06-11)
The accumulation of RCY1 protein, which is encoded by RESISTANCE TO CMV(Y) (RCY1), a CC-NB-LRR class R-gene, is tightly correlated with the strength of the resistance to a yellow strain of Cucumber mosaic virus [CMV(Y)] in Arabidopsis thaliana. In order
Azadeh Nilghaz et al.
Lab on a chip, 12(1), 209-218 (2011-11-18)
This paper describes the fabrication of microfluidic cloth-based analytical devices (μCADs) using a simple wax patterning method on cotton cloth for performing colorimetric bioassays. Commercial cotton cloth fabric is proposed as a new inexpensive, lightweight, and flexible platform for fabricating
Franziska Kaplan et al.
Micron (Oxford, England : 1993), 44, 317-330 (2012-09-11)
The osmotic potential and effects of plasmolysis on photosynthetic oxygen evolution and chlorophyll fluorescence were studied in two Arctic Zygnema sp. (strain B, strain G) and two Antarctic Zygnema sp. (strain E, strain D). Antarctic strain D was newly characterized
Dries Vandamme et al.
Bioresource technology, 105, 114-119 (2011-12-21)
Microalgae hold great potential as a feedstock for biofuels or bulk protein or treatment of wastewater or flue gas. Realising these applications will require the development of a cost-efficient harvesting technology. Here, we explore the potential of flocculation induced by
Sandra Nilsson et al.
PloS one, 7(11), e46980-e46980 (2012-11-28)
During the intraerythrocytic development of Plasmodium falciparum, the malaria parasite remodels the host cell cytosol by inducing membranous structures termed Maurer's clefts and inserting parasite proteins into the red blood cell cytoskeleton and plasma membrane. Pf332 is the largest known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service