Skip to Content
Merck
All Photos(1)

Key Documents

78348

Supelco

Sodium acetate/Sodium hydroxide eluent

10 mM CH3COONa in H2O, 100 mM NaOH in H2O, IC eluent for Metrosep Carb 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

concentration

10 mM CH3COONa in H2O
100 mM NaOH in H2O

technique(s)

ion chromatography: suitable

suitability

application tested

General description

This eluent concentrate is application tested. Information about the exact contents and expiry date is described in the certificate.

Application

Metrohm IC application note AN-P-072:Carbohydrates in orange juice applying pulsed amperometric detection.

Metrohm IC application note AN-P-067: Sorbitol andsucrose in soap applying IC amperometric detection.

Metrohm IC application note AN-P-065: Sugar and sugar alcohols in addition to sucrose and cellobiose.

Metrohm IC application note AN-P-062: Sugar and sugar alcohols in an apple beverage with pulsed amperometric detection.

Linkage

Visit the IC Portal to learn more

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In this article we demonstrate how well ion chromatography is suited as an analytical technique for sugar analysis e.g. in apple juice, milk, and dust samples.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service