Skip to Content
Merck
All Photos(1)

Documents

71943

Sigma-Aldrich

Sodium salicylate

tested according to Ph. Eur.

Synonym(s):

Natrii salicylas, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COONa
CAS Number:
Molecular Weight:
160.10
Beilstein:
3732792
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

crystals

color

white

useful pH range

6.85 (26.3 °C)

mp

>300 °C (lit.)

solubility

water: 575.7 g/L at 25 °C (77 °F)

application(s)

pharmaceutical (small molecule)

SMILES string

[Na+].Oc1ccccc1C([O-])=O

InChI

1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ABBQHOQBGMUPJH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

208.9 °F - Pensky-Martens closed cup

Flash Point(C)

98.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kui Wang et al.
Journal of medicinal chemistry, 52(20), 6402-6412 (2009-10-16)
Viologens are showing an increasing number of scientific and technical applications in addition to their use as herbicides. However, their high toxicity poses considerable risks to human health, society, and the environment. In this context, we propose a new therapeutic
Yanguang Cao et al.
The Journal of pharmacology and experimental therapeutics, 339(3), 896-904 (2011-09-10)
Type 2 diabetes mellitus (T2DM) arises owing to insulin resistance and β-cell dysfunction. Chronic inflammation is widely identified as a cause of T2DM. The Goto-Kakizaki (GK) rat is a spontaneous rodent model for T2DM with chronic inflammation. The purpose of
Rainer Amann et al.
European journal of pharmacology, 447(1), 1-9 (2002-07-11)
Aspirin (acetylsalicylic acid) is one of the most widely used drugs worldwide. It acetylates cyclooxygenases thereby irreversibly blocking the conversion of arachidonic acid to prostanoids. Biotransformation of aspirin yields salicylate, a compound that possesses similar anti-inflammatory potency as aspirin but
Yan-Yan Su et al.
PloS one, 7(10), e46969-e46969 (2012-10-17)
Sodium salicylate (NaSal), an aspirin metabolite, can cause tinnitus in animals and human subjects. To explore neural mechanisms underlying salicylate-induced tinnitus, we examined effects of NaSal on neural activities of the medial geniculate body (MGB), an auditory thalamic nucleus that
Yutaka Sumino et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 85(4 Pt 2), 046307-046307 (2012-06-12)
The dynamics of a viscoelastic Maxwell fluid is studied in a partially filled cylinder rotating around a horizontal axis. At low rotational velocity, the fluid behaves in the same manner as a viscous fluid. A thin fluid film is pulled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service