567630
Sodium Salicylate
A cell-permeable, non-steroidal anti-inflammatory agent that interferes with TNF-induced NF-κB activation by inhibiting phosphorylation and subsequent degradation of the IκB-α protein.
Synonym(s):
Sodium Salicylate, Salicylic Acid, Na, NaSal
Select a Size
€57.40
Estimated to ship on30 March 2025
Select a Size
About This Item
€57.40
Estimated to ship on30 March 2025
Recommended Products
Quality Level
Assay
≥98% (titration)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
color
off-white
solubility
water: 50 mg/mL
ethanol: soluble
shipped in
ambient
storage temp.
10-30°C
SMILES string
[Na+].[O-]C(=O)c1c(cccc1)O
InChI
1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1
InChI key
ABBQHOQBGMUPJH-UHFFFAOYSA-M
General description
Biochem/physiol Actions
TNF-induced activation of p42 (ERK2) and p44 (ERK1) MAP kinases
Packaging
Warning
Reconstitution
Other Notes
Mitchell, J.A., et al. 1997. Mol. Pharmacol. 51, 907.
Schwenger, P., et al. 1997. Proc. Natl. Acad. Sci. USA 94, 2869.
Farivar, R.S. and Brecher, P. 1996. J. Biol. Chem. 271, 31585.
Schwenger, P., et al. 1996. J. Biol. Chem. 271, 8089.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
208.9 °F - Pensky-Martens closed cup
Flash Point(C)
98.3 °C - Pensky-Martens closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service