Skip to Content
Merck
All Photos(1)

Key Documents

69527

Millipore

Modified Giolitti and Cantoni Broth (ISO)

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Giolitti Cantoni Broth, Modified (ISO)

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€119.00

€119.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€119.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

€119.00


Estimated to ship on28 May 2025


Request a Bulk Order

Agency

according to ISO 6888-3:2003

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

beef extract, 5.0 g/L
casein enzymic hydrolysate, 10.0 g/L
glycine, 1.2 g/L
lithium chloride, 5.0 g/L
mannitol, 20.0 g/L
sodium chloride, 5.0 g/L
sodium pyruvate, 3.0 g/L
yeast extract, 5.0 KBE/mL

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.9±0.2 (25 °C)

application(s)

environmental
food and beverages
sample preparation

microbiology

suitability

Staphylococcus spp.

Application

Giolitti Cantoni Broth Base with addition of potassium tellurite is used for selective enrichment of Staphylococcus aureus from foods.

Preparation Note

Suspend 54.2 grams in 1000 ml distilled water. Warm gently to dissolve the medium completely. Dispense 19 ml amounts in 20mm x 200mm test tubes. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool rapidly to room temperature and aseptically add 0.3 ml of 3.5% Potassium Tellurite Solution to each tube. Add 0.03 ml for testing meat and meat products. Mix well before use.Caution : Lithium chloride is harmful. Avoid bodily contact and inhalation of vapours. On contact with skin, wash with plenty of water immediately

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service