Skip to Content
Merck
All Photos(2)

Key Documents

1.10426

Millipore

Chromocult® Coliform Agar

ISO 9308-1: 2014, For the simultaneous detection and enumeration of coliform bacteria and E. coli

Synonym(s):

Coliform Agar, Chromogenic E. coli Agar, Chromogenic Coliform Agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€806.00

€806.00


Check Cart for Availability

Request more information

Select a Size

Change View
500 G
€806.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106213
NACRES:
NA.74

€806.00


Check Cart for Availability

Request more information

Agency

AOAC® RI (approved as alternative method for processed foods)
EPA
ISO 9308-1: 2014

Quality Level

sterility

non-sterile

form

granular

manufacturer/tradename

ChromoCult®

technique(s)

microbiological culture: suitable

pH

6.6-7.0 (25 °C, 26.5 g/L in H2O)

solubility

26.5 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Escherichia coli
coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Chromocult® Coliform Agar is a granulated, less selective, and differential chromogenic culture medium intended for microbiological analysis of food and water. This medium saves time by enabling the simultaneous detection, differentiation, and enumeration of E. coli and coliforms from drinking water and processed food matrices within 24 hours. This agar is the only chromogenic culture medium which was used for validation to prepare the new ISO 9308-1 (2014) version. The medium contains Tergitol 7 as an inhibitor of Gram-positive bacteria without any impact on the growth of the targeted coliforms/ E. coli. Chromocult® Coliform Agar is, therefore, the ideal medium for the detection of coliforms/ E. coli in drinking water and processed foods. It facilitates and speeds up your processes.

Application

Chromocult® Coliform Agar is used for the simultaneous detection of coliform bacteria and E. coli in drinking water, disinfected pool water, finished water from treatment plants and processed food samples.

Analysis Note

Appearance (clearness): slightly turbid to turbid
Appearance (colour): yellowish to yellowish-brown
pH-value (25 °C): 6.6 - 7.0
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)):
Inoculum on reference medium (Escherichia coli ATCC 8739 (WDCM 00012)):
Inoculum on reference medium (Enterobacter aerogenes ATCC 13048):
Inoculum on reference medium (Citrobacter freundii ATCC 43864 (WDCM 00006)):
Colony count (Escherichia coli ATCC 25922 (WDCM 00013)):
Colony count (Escherichia coli ATCC 8739 (WDCM 00012)):
Colony count (Enterobacter aerogenes ATCC 13048):
Colony count (Citrobacter freundii ATCC 43864 (WDCM 00006)):
Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 70 %
Recovery on test medium (Escherichia coli ATCC 8739 (WDCM 00012)): ≥ 70 %
Recovery on test medium (Enterobacter aerogenes ATCC 13048): ≥ 70 %
Recovery on test medium (Citrobacter freundii ATCC 43864 (WDCM 00006)): ≥ 70 %
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): partial inhibition
Growth (Pseudomonas aeruginosa ATCC 10145 (WDCM 00024)): no limit
Colonies (Escherichia coli ATCC 25922 (WDCM 00013)): dark-blue to violet
Colonies (Escherichia coli ATCC 8739 (WDCM 00012)): dark-blue to violet
Colonies (Enterobacter aerogenes ATCC 13048 (WDCM 00175)): pink to red
Colonies (Citrobacter freundii ATCC 43864 (WDCM 00006)): pink to red
Colonies (Pseudomonas aeruginosa ATCC 10145 (WDCM 00024)): colourless
Incubation: 21 ± 3 hours at 36 ± 2 °C
A recovery rate of 70 % is equivalent to a productivity value of 0.7.
The indicated colony counts result from the sum of a triple determination.
Reference media: Tryptic Soy Agar

Legal Information

AOAC is a registered trademark of AOAC International
CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Related Content

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Questions

1–2 of 2 Questions  
  1. Wie wird der Agar aus dem Pulver hergestellt. Muss autoklaviert werden?

    1 answer
    1. Preparation instructions are as follow:

      Dissolve 26.5 g in 1 liter of purified water. Heat in boiling water and agitate frequently until completely dissolved. Do not autoclave, do not overheat!

      Cool the medium to 45 - 50 °C and pour plates. According to EN ISO 9308-1 dispense in Petri dishes to a depth of at least 4 mm (approximately 18 ml in a 90 mm plate).

      For using by poured plate method cool the medium to 44 - 47 °C in a water bath before use (see EN ISO 11133).

      The prepared medium is slightly turbid to turbid and yellowish. The pH value at 25 °C is in the range of 6.6-7.0

      There should be no visible moisture on the plates before use. When moisture is present, the plates should be dried for the minimum time required to remove visible moisture, following the procedure as described by EN ISO 11133.

      Preparation and addition of optional Cefsulodin solution (US-EPA approved method): Dissolve 5 mg of Cefsulodin in 2 ml of purified water and sterilize by membrane filtration (0.2 μm nominal pore size). Aseptically add the solution (2 ml per 500 ml of medium) to 500 ml of liquefied medium (45 - 50 °C). Pour plates as described above.

      Helpful?

  2. What precautions do you need to take to produce chromocult Coliformes agar, since it cannot be overheated or autoclaved? What are the recommendations? Sterile water? Sterile glassware?

    1 answer
    1. Ideally, a boiling hot water bath would be best. However, you may also use a hot plate with a magnetic stirrer to prepare the medium. Using a magnetic stir bar, thoroughly mix the powdered media with purified water until no clumps are visible. Use a container larger than the volume being prepared (for example - 1L Erlinmeyer flask to prepare 500 mL media). Heat media to 100°C, which will be the boiling temperature. Boiling will melt the agar, but bubbling will occur. Closely monitor to prevent boiling over. Media is sufficiently heated when the appearance transitions from cloudy/hazy to transparent. Allow media to cool to 45 - 50°C and pour aseptically onto plates. For larger volumes, the media can be dispensed by using a peristaltic pump, Sterile plates, utensils, and/or tubing may be used during the preparation and filling process.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service