Skip to Content
Merck
All Photos(1)

Key Documents

66003BC

Millipore

SimPlate® Total Plate Count

BioControl, Quantitative results for total aerobic bacteria from food and environmental samples in just 24 hours

Synonym(s):

TPC, Single

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€144.00
1 SET
€448.00

€144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
€144.00
1 SET
€448.00

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
material:
plastic vial (containing dehydrated culture media)
feature:
fluorescent indicator

€144.00


Check Cart for Availability

Request a Bulk Order

material

plastic vial (containing dehydrated culture media)

size

500 tests

Agency

AOAC® PTM

form

dry powder

shelf life

3‑18 mo.

feature

fluorescent indicator

packaging

container of 100

manufacturer/tradename

BioControl Systems 66003-100
BioControl Systems 66003-500

storage condition

dry media

technique(s)

cell counting: suitable (total aerobic microorganisms)

application(s)

food and beverages

compatibility

for use with SimPlate (65009BC)

storage temp.

room temp

General description

SimPlate for Total Plate Count (TPC) method is used for the detection and quantification of the total aerobic microorganisms in food. The medium/sample mixture is dispensed into a SimPlate device and incubated for 24-28 h. The total aerobic plate count is determined by counting the fluorescent wells and referring to the SimPlate Conversion Table. The SimPlate device is packaged separately.

Features and Benefits

  • Rapid Enumeration of Total Aerobic Bacteria
  • Easy-to-Read Results
  • While other methods require duplicate plating of samples, SimPlate has been validated to provide equivalent results with just a single plate. Combined with the reduction of necessary sample dilutions, made possible by the broad counting range of the plate (1-738), SimPlate reduces the number of tests required for each sample, helping to reduce overall testing costs.

Components

2 packages of 50 single-test vials (66003-100), or 1 package of 50 multi-test (10 tests/vial) vials (66003-500)

Testing Method

Binary Detection Technology®

Other Notes

Required for use: SimPlate Device (65009BC), Recommended for use: Supplement A (63011BC)

Legal Information

AOAC is a registered trademark of AOAC International
Binary Detection Technology is a registered trademark of Merck KGaA, Darmstadt, Germany
SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • SimPlate Total Plate Count (TPC) Single Dose Fill, Irradiated, 100 test

  • SimPlate Total Plate Count (TPC) Multi Dose Fill, Irradiated, 500 test

required but not provided

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service