Skip to Content
Merck
All Photos(1)

Key Documents

63010BC

Millipore

Supplement D, SimPlate®

Supplement D, SimPlate®

BioControl, For use with SimPlate Yeast & Mold Color Indicator

Synonym(s):

Supplement

Sign Into View Organizational & Contract Pricing

Select a Size

40 ML
€26.10

€26.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
40 ML
€26.10

About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

€26.10


Check Cart for Availability

Request a Bulk Order

material

plastic vial (containing dehydrated culture media)

form

liquid

shelf life

6‑36 mo.

packaging

bottle of 1 40 mL

manufacturer/tradename

BioControl Systems 63010-200

storage condition

protect from light

application(s)

food and beverages

compatibility

for use with SimPlate (66007BC)

storage temp.

2-8°C

suitability

molds
yeasts

General description

Supplement D is an additional reagent for use in the SimPlate® yeast & mold. SimPlate® Binary Detection Technology combines proprietary media and a patented plating device to provide fast and accurate quantitative results for key groups of indicator organisms. Defined substrates incorporated into the media are utilized to detect multiple enzymes and redox reactions associated with the target organisms resulting in chromogenic or fluorescent changes to the media, providing easy-to-read results. Supplement D is added to prevent the mold overgrowth

Application

Supplement D is added to SimPlate® trays to prevent mold over-growth in media on samples with spreading molds without the need for optimization of reaction and cycle conditions.

Components

1 bottle, approx. 40 mL (adequate for 200 tests)

Other Notes

For use with SimPlate Yeast & Mold CI (66007BC)

Legal Information

SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. Eau Claire, WI
    • Reviews 2
    • Vote 1
    5 out of 5 stars.

    Supplement D

    Takes the place of USP fluid D for Simplate sample preparations.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service