Skip to Content
Merck
All Photos(1)

Key Documents

66009BC

Millipore

SimPlate® Enterobacteriaceae CI

BioControl, Quantitative results for Enterobacteriaceae from food and environmental samples in just 24 hours

Synonym(s):

Entero-CI (Color Indicator)

Sign Into View Organizational & Contract Pricing

Select a Size

100 EACH
€235.00

€235.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 EACH
€235.00

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
material:
plastic vial (containing dehydrated culture media)
suitability:
enterobacteriaceae
feature:
fluorescent indicator

€235.00


Check Cart for Availability

Request a Bulk Order

material

plastic vial (containing dehydrated culture media)

size

100 tests

form

dry powder

shelf life

6‑18 mo.

feature

fluorescent indicator

packaging

container of 100

manufacturer/tradename

BioControl Systems 66009-100

storage condition

dry media

application(s)

food and beverages

compatibility

for use with SimPlate (65009BC)

storage temp.

room temp

suitability

enterobacteriaceae

Related Categories

General description

SimPlate for Total Enterobacteriaceae Color Indicator (Entero-CI) method is used for the detection and quantification of the total Enterobacteriaceae populations. It is based on Binary Detection Technology (BDT) which equates the presence of total Enterobacteriaceae to the presence of a color change in the medium. The medium/sample mixture is dispensed into a SimPlate device and incubated for 24-28 h. The total Enterobacteriaceae count is determined by counting the wells with changed color and referring to the SimPlate Conversion Table. Colored wells indicate the presence of total Enterobacteriaceae.The SimPlate device is packaged separately.

Features and Benefits

  • SimPlate Enterobacteriaceae utilizes proprietary Binary Detection Technology® to provide quantitative results for members of the Enterobacteriaceae family from food and environmental samples in just 24 hours.
  • Proprietary chromogenic media and patented plating device with isolation wells combine to produce easily counted results. Positive wells are clearly identified based on a visual color change.
  • While other plating methods require duplicate plating of samples, SimPlate provides equivalent results with just a single plate. Combined with the reduction of necessary sample dilutions, made possible by the broad counting range of the plate (1-738), SimPlate reduces the volume of media and number of tests required for each sample, helping to reduce overall testing costs.

Components

2 packages of 50 vials (one test per vial)

Testing Method

Binary Detection Technology®

Other Notes

Required for use: SimPlate Device (65009BC)

Legal Information

Binary Detection Technology is a registered trademark of Merck KGaA, Darmstadt, Germany
SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • Entero-CI Single-dose fill

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service