Skip to Content
Merck

Skip To

60741

Silica gel Inorganic Sorbent

high-purity grade, 70-230 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 kg
Check Cart for Availability
€ 137,00
5 kg
Check Cart for Availability
€ 706,00
25 kg
Check Cart for Availability
€ 2.420,00

About This Item

CAS Number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
SB.52
EC Number:
231-545-4
MDL number:

€ 137,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Silica gel, high-purity grade, average pore size 60 Å (52-73 Å), 70-230 mesh, 63-200 μm, for column chromatography

grade

high-purity grade

Quality Segment

description

activity acc. to Brockmann and Schodder 2 - 3

form

powder or crystals

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

surface area

470-530 m2/g

matrix

Silica

matrix active group

silica

particle size

63-200 μm, 70-230 mesh

pore size

0.7-0.85 cm3/g pore volume, 60 Å average pore size (52-73 Å)

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction

Application

Silica gel may be used in separation of single-walled carbon nanotube (SWNT) by simple filtration.[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
6073760738288519
matrix active group

silica

matrix active group

silica

matrix active group

silica

matrix active group

-

particle size

63-200 μm, 70-230 mesh

particle size

230-400 mesh, 40-63 μm

particle size

220-440 mesh, 35-75 μm

particle size

5-25 μm

separation technique

hydrophilic interaction (HILIC)

separation technique

hydrophilic interaction (HILIC)

separation technique

hydrophilic interaction (HILIC)

separation technique

-

matrix

Silica

matrix

Silica

matrix

Silica

matrix

-

pore size

0.7-0.85 cm3/g pore volume, 60 Å average pore size (52-73 Å)

pore size

0.8 cm3/g pore volume, 60 Å

pore size

0.8 cm3/g pore volume, 60 Å

pore size

0.75 cm3/g pore volume, 60 Å medium pore diameter, 60 Å pore size

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

thin layer chromatography (TLC): suitable


Still not finding the right product?

Explore all of our products under Silica gel


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Christopher A Dyke et al.
Journal of the American Chemical Society, 127(12), 4497-4509 (2005-03-24)
In this report, procedures are discussed for the enrichment of single-walled carbon nanotube (SWNT) types by simple filtration of the functionalized SWNTs through silica gel. This separation uses nanotube sidewall functionalization employing two different strategies. In the first approach, a
A simple method for production of pure silica from rice hull ash
Kalapathy, U., A. Proctor, and J. Shultz.
Bioresource Technology, 73, 257-262 (2000)
Silica gel supported zirconocene dichloride/methylalumoxane catalysts for ethylene polymerization: Effects of heterogenation on activity, polymer microstructure and product morphology.
Janiak, Christoph, and Bernhard Rieger.
Angew. Makromol. Chem., 215, 47-57 (1994)



Global Trade Item Number

SKUGTIN
15073-5G04061832463087
60741-1KG04061838550453
60741-25KG04061832912547
60741-5KG04061838550460

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters