Skip to Content
Merck

Skip To

46005

Fenclorim

PESTANAL®, analytical standard

Synonym(s):

4,6-Dichloro-2-phenylpyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H6Cl2N2
CAS Number:
Molecular Weight:
225.07
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
142293
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1cc(Cl)nc(n1)-c2ccccc2

InChI

1S/C10H6Cl2N2/c11-8-6-9(12)14-10(13-8)7-4-2-1-3-5-7/h1-6H

InChI key

NRFQZTCQAYEXEE-UHFFFAOYSA-N

General description

Fenclorim is a herbicide saferner belonging to the pyrimidine class, used for the protection of crops from seedling damage. It mainly acts by metabolizing pretilachlor in direct seeded rice.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Fenclorim may be used as an analytical reference standard for the quantification of the analyte in environmental samples and food crops using different chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



The Pesticide Encyclopedia (2014)
Development and validation of a multiresidue method for determination of 82 pesticides in water using GC
Mamun R.IM, et al.
Journal of Separation Science, 32(4), 559-574 (2009)
Quantification of pesticides in food crops using QuEChERS approaches and GC-MS/MS
Cho J, et al.
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment, 33(12), 1803-1816 (2016)



Global Trade Item Number

SKUGTIN
115931002504022536162195
46005-250MG04061832343679

Questions

Reviews

No rating value

Active Filters