Skip to Content
Merck
All Photos(2)

Documents

45481

Supelco

Etrimfos

PESTANAL®, analytical standard

Synonym(s):

O-(6-Ethoxy-2-ethyl-4-pyrimidinyl) O,O-dimethyl thiophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N2O4PS
CAS Number:
Molecular Weight:
292.29
Beilstein:
7136389
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

technical mixture

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOc1cc(OP(=S)(OC)OC)nc(CC)n1

InChI

1S/C10H17N2O4PS/c1-5-8-11-9(15-6-2)7-10(12-8)16-17(18,13-3)14-4/h7H,5-6H2,1-4H3

InChI key

FGIWFCGDPUIBEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

T Cairns et al.
Journal - Association of Official Analytical Chemists, 70(5), 858-862 (1987-09-01)
The production of a protonated molecular ion, MH+, for a pesticide under investigation is often preferred for primary identification purposes. However, the lack of fragment ions under such chemical ionization conditions can place the burden of proof on a single
Merve Fırat et al.
Environmental monitoring and assessment, 191(10), 619-619 (2019-09-08)
Presented in this study is a simple but efficient switchable polarity solvent microextraction strategy for etrimfos preconcentration from water and food samples for quantification by gas chromatography mass spectrometry. Repeatability of the extraction process and instrumental measurements were enhanced by
[Determination of the ekamet content of vegetables and apples by thin-layer chromatography].
V S Shustrov et al.
Voprosy pitaniia, (3)(3), 70-71 (1984-05-01)
Bartosz Sell et al.
Journal of analytical toxicology, 42(3), 183-191 (2017-12-02)
Reported here is a new analytical multiclass method based on QuEChERS technique, which has proven to be effective in diagnosing fatal poisoning cases in animals. This method has been developed for the determination of analytes in liver samples comprising rodenticides
Marek Łukasz Roszko et al.
PloS one, 11(12), e0167487-e0167487 (2016-12-03)
Chemical plant protection is a commonly discussed factor potentially responsible for decline in pollinators and other beneficial insect populations. Various groups of chemicals including persistent organic pollutants could impact a bee colony's welfare and are reported to be present in

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Chromatograms

application for LC-MS, application for SPE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service