Skip to Content
Merck
All Photos(1)

Key Documents

31644

Supelco

Bromuconazole

PESTANAL®, analytical standard, mixture of diastereomers

Synonym(s):

1-[[4-Bromo-2-(2,4-dichlorophenyl)tetrahydro-2-furanyl]methyl]- 1H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€180.00

€180.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€180.00

About This Item

Empirical Formula (Hill Notation):
C13H12BrCl2N3O
CAS Number:
Molecular Weight:
377.06
Beilstein:
8393967
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€180.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

description

mixture of diastereomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(c(Cl)c1)C2(CC(Br)CO2)Cn3cncn3

InChI

1S/C13H12BrCl2N3O/c14-9-4-13(20-5-9,6-19-8-17-7-18-19)11-2-1-10(15)3-12(11)16/h1-3,7-9H,4-6H2

InChI key

HJJVPARKXDDIQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Bromuconazol may be used as a reference standard for the determination of the analyte:
  • In food and water samples using liquid chromatography/time-of-flight mass spectrometry.[1]
  • In orange juice using ultra high performance liquid chromatography coupled to tandem mass spectrometry (UHPLC–MS/MS).[2]
  • In fruit jams using micro flow liquid chromatography–tandem mass spectrometry (μLC–MS/MS).[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation and application of micro flow liquid chromatography- tandem mass spectrometry for the determination of pesticide residues in fruit jams.
Reichert B, et al.
Talanta, 134(1), 415- 424 (2015)
Optimization of a QuEChERS based method by means of central composite design for pesticide multiresidue determination in orange juice by UHPLC- MS/MS.
Rizzetti TM, et al.
Food Chemistry, 196(1), 25- 33 (2016)
Multi-residue method for the analysis of 101 pesticides and their degradates in food and water samples by liquid chromatography/time-of-flight mass spectrometry.
Ferrer I, Thurman EM
Journal of Chromatography A, 1175(1), 24-37 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service