Skip to Content
Merck
All Photos(2)

Key Documents

33395

Supelco

Methamidophos

PESTANAL®, analytical standard

Synonym(s):

O,S-Dimethyl phosphoramidothioate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€65.10

€65.10


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€65.10

About This Item

Empirical Formula (Hill Notation):
C2H8NO2PS
CAS Number:
Molecular Weight:
141.13
Beilstein:
8137714
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€65.10


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water (Karl Fischer)

mp

42-45 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

COP(N)(=O)SC

InChI

1S/C2H8NO2PS/c1-5-6(3,4)7-2/h1-2H3,(H2,3,4)

InChI key

NNKVPIKMPCQWCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methamidophos is a broad spectrum organophosphate pesticide used for controlling a variety of plant and soil pests in agricultural crops.[1]

Application

Methamidophos may be used as a reference standard for the determination of the analyte in human serum samples using gas chromatography with mass spectrometry (GC-MS).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

413.6 °F - closed cup

Flash Point(C)

212 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Impacts of methamidophos on the biochemical, catabolic, and genetic characteristics of soil microbial communities.
Wang CM, et al.
Critical reviews in environmental science and technology, 40(3), 778-788 (2008)
Simultaneous analysis of acephate and methamidophos in human serum by improved extraction and GC-MS.
Adachi N, et al.
Forensic Toxicology, 26(2), 76-76 (2008)
Teresa Rodríguez et al.
Occupational and environmental medicine, 69(2), 119-125 (2011-07-05)
This study assessed pesticide exposure of children in rural Nicaragua in relation to parental pesticide use, from around conception to current school age, as part of an epidemiological evaluation of neurodevelopment effects. We included 132 children whose parents were subsistence
Carla S Lima et al.
Neurotoxicology, 32(6), 718-724 (2011-08-30)
Epidemiologic studies describe a potential risk of depression and suicide in farm workers exposed to organophosphates (OPs). In a previous study we observed an increase in depressive-like behavior in adult mice exposed to the OP pesticide methamidophos. Considering the association
Guilherme L Emerick et al.
Toxicology, 292(2-3), 145-150 (2011-12-27)
The current Organisation for Economic Co-operation and Development (OECD) guidelines for evaluating organophosphorus-induced delayed neuropathy (OPIDN) require the observation of dosed animals over several days and the sacrifice of 48 hens. Adhering to these protocols in tests with enantiomers is

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service