Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
analytical standard
Quality Segment
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
CN(C)C(=O)NC1CCCCCCC1
InChI
1S/C11H22N2O/c1-13(2)11(14)12-10-8-6-4-3-5-7-9-10/h10H,3-9H2,1-2H3,(H,12,14)
InChI key
DQZCVNGCTZLGAQ-UHFFFAOYSA-N
Application
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| format neat | format neat | format neat | format neat |
| technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable |
| grade analytical standard | grade analytical standard | grade analytical standard | grade analytical standard |
| product line PESTANAL® | product line PESTANAL® | product line PESTANAL® | product line PESTANAL® |
| application(s) agriculture | application(s) agriculture | application(s) agriculture | application(s) agriculture |
| shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




