Skip to Content
Merck
All Photos(1)

Documents

41293

Supelco

Acid Violet 49

analytical standard

Synonym(s):

[4-[[4-(Dimethylamino)phenyl][4-[ethyl(3-sulphonatobenzyl)amino]phenyl]methylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(3-sulphonatobenzyl)ammonium sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H40N3NaO6S2
CAS Number:
Molecular Weight:
733.87
Colour Index Number:
42640
EC Number:
UNSPSC Code:
85151701

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

InChI

1S/C39H41N3O6S2.Na/c1-5-41(27-29-9-7-11-37(25-29)49(43,44)45)35-21-15-32(16-22-35)39(31-13-19-34(20-14-31)40(3)4)33-17-23-36(24-18-33)42(6-2)28-30-10-8-12-38(26-30)50(46,47)48;/h7-26H,5-6,27-28H2,1-4H3,(H-,43,44,45,46,47,48);/q;+1/p-1

InChI key

AXMCIYLNKNGNOT-UHFFFAOYSA-M

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiro Iijima et al.
Journal of clinical laboratory analysis, 17(5), 147-154 (2003-08-26)
We have developed a new method for measuring total urinary protein using acid violet 6B (AV6B) pigment. Nine purified components of human urinary proteins and urine samples collected randomly from 123 diabetic outpatients were used. There were 62, 36, and
Weixuan Lin et al.
Se pu = Chinese journal of chromatography, 30(5), 527-532 (2012-09-01)
A method of high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) has been established for the simultaneous determination of six forbidden colorants including Sudan IV, Acid Violet 49, Sudan Blue 2, Solvent Red 49, Basic Violet 1 and Pigment Orange 5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service