Skip to Content
Merck
All Photos(2)

Key Documents

195219

Sigma-Aldrich

Mordant Blue 9

Dye content 50 %

Synonym(s):

6-(5-Chloro-2-hydroxy-3-sulfophenylazo)-5-hydroxy-1-naphthalenesulfonic acid disodium salt, 6-(5-Chloro-2-hydroxy-4-sulfonphenylazo)-5-hydroxy-1-naphthalene-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€59.50

€59.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€59.50

About This Item

Linear Formula:
ClC6H2(OH)(SO3Na)N=NC10H5(OH)SO3Na
CAS Number:
Molecular Weight:
502.81
Colour Index Number:
14855
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€59.50


Check Cart for Availability

Request a Bulk Order

form

solid

composition

Dye content, 50%

solubility

H2O: 1 mg/mL

λmax

516 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1c(cc(Cl)cc1S([O-])(=O)=O)\N=N\c2ccc3c(cccc3c2O)S([O-])(=O)=O

InChI

1S/C16H11ClN2O8S2.2Na/c17-8-6-12(16(21)14(7-8)29(25,26)27)19-18-11-5-4-9-10(15(11)20)2-1-3-13(9)28(22,23)24;;/h1-7,20-21H,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2/b19-18+;;

InChI key

UVDODHCPLWXDSI-BUFQOAPZSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service