Skip to Content
Merck

Skip To

44565

Tropaeolin O sodium salt

indicator grade

Synonym(s):

4-([2,4-Dihydroxyphenyl]azo)benzenesulfonic acid sodium salt, Acid Orange 6, Chrysoin, Resorcinol yellow

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
€ 93,40

About This Item

Empirical Formula (Hill Notation):
C12H9N2O5SNa
CAS Number:
Molecular Weight:
316.27
UNSPSC Code:
41116105
NACRES:
NA.21
MDL number:
Colour Index Number:
14270
Beilstein/REAXYS Number:
4173410

€ 93,40


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

indicator grade

composition

sulfated ash, 40-60%

technique(s)

UV/Vis spectroscopy: suitable, titration: suitable

loss

≤13.0% loss on drying

pH

11-13

λmax

488-492 nm in 0.1 M NaOH, ≥500 at 1% in 0.1 M NaOH (1 cm)

SMILES string

[Na+].Oc1ccc(\N=N\c2ccc(cc2)S([O-])(=O)=O)c(O)c1

InChI

1S/C12H10N2O5S.Na/c15-9-3-6-11(12(16)7-9)14-13-8-1-4-10(5-2-8)20(17,18)19;/h1-7,15-16H,(H,17,18,19);/q;+1/p-1/b14-13+;

InChI key

COEZWFYORILMOM-IERUDJENSA-M


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.5926719523569143
technique(s)

UV/Vis spectroscopy: suitable, titration: suitable

technique(s)

-

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

indicator grade

grade

-

grade

-

grade

analytical standard

pH

11-13

pH

10 (20 °C, 10 g/L in H2O)

pH

-

pH

-

loss

≤13.0% loss on drying

loss

-

loss

-

loss

-

composition

sulfated ash, 40-60%

composition

-

composition

Dye content, ≥85%

composition

-

λmax

488-492 nm in 0.1 M NaOH

λmax

-

λmax

483 nm

λmax

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters