Skip to Content
Merck

Skip To

41008

Ethylenediamine

analytical standard

Synonym(s):

1,2-Diaminoethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Check Cart for Availability
€ 29,20

About This Item

Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
NACRES:
NA.24
PubChem Substance ID:
eCl@ss:
39030201
UNSPSC Code:
41116107
EC Number:
203-468-6
MDL number:
Beilstein/REAXYS Number:
605263

€ 29,20


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

2.07 (vs air)

vapor pressure

10 mmHg ( 20 °C)

assay

≥99.8% (GC)

autoignition temp.

716 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

16 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.456, n20/D 1.4565 (lit.)

bp

118 °C (lit.)

mp

8.5 °C (lit.)

solubility

H2O: soluble at 

density

0.899 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

NCCN

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2

InChI key

PIICEJLVQHRZGT-UHFFFAOYSA-N

General description

This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E164903550391085
application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

format

neat

format

-

format

-

format

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

grade

analytical standard

grade

-

grade

-

grade

-

assay

≥99.8% (GC)

assay

98.6% (Gew)

assay

≥99.5% (GC)

assay

≥99.5%


Still not finding the right product?

Explore all of our products under Ethylenediamine


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

100.4 °F - closed cup

flash_point_c

38 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

107-15-3

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters