Skip to Content
Merck

Skip To

39396

Casein Yeast Peptone

suitable for microbiology

Synonym(s):

Casein Peptone tryptic digest / Yeast Extract (4:1), Tryptone / Yeast Extract (4:1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 g
Check Cart for Availability
€ 138,00

About This Item

NACRES:
NA.85
UNSPSC Code:
41106212

€ 138,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

bovine milk, yeast

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥2.5-5.0% , total nitrogen (N), ≥12%

packaging

pkg of 500 g

storage condition

(Tightly closed. Dry.)

impurities

≤15% ash

loss

<10% loss on drying

pH

7.2±0.5 (2% in H2O)

solubility

H2O: 2%, clear, yellow

application(s)

food and beverages, microbiology

General description

Casein Yeast Peptone is a common peptone mixture often used in microbiology for the growth of bacteria and fungi. It is a complex mixture that provides a rich source of nutrients necessary for the growth and metabolism of microorganisms.The main components of Casein Yeast Peptone are casein peptone and yeast extract. Casein peptone is a milk protein that is hydrolyzed into smaller peptides, providing a source of amino acids for microbial growth. Yeast extract is derived from the autolysis of yeast cells, and contains a range of nutrients such as vitamins, minerals, and amino acids.

Application

Casein Yeast Peptone creates a nutrient-rich and versatile medium suitable for a wide range of microorganisms. This medium supports the growth of fastidious organisms and is commonly used in microbial culture, isolation, and identification techniques, as well as in the production of enzymes, antibiotics, and various biotechnological processes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
6104470172T9410
biological source

bovine milk, yeast

biological source

bovine milk

biological source

bovine milk

biological source

bovine milk

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

food and beverages, microbiology

application(s)

food and beverages
microbiology

application(s)

food and beverages
microbiology

application(s)

microbiology

form

powder

form

powder

form

powder

form

powder

pH

7.2±0.5 (2% in H2O)

pH

6.2-7.2 (25 °C)

pH

6.7±0.5 (25 °C, 2% in H2O)

pH

6.6-7.4

solubility

H2O: 2%, clear, yellow

solubility

H2O: 2%, clear, faintly yellow to brownish-yellow

solubility

H2O: 2%, clear, colorless to faintly brownish-yellow

solubility

-


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.






Global Trade Item Number

SKUGTIN
529982-25G04061837074004
39396-500G-F04061833661635

Questions

  1. Cab it be used for edible product development?

    1 answer
    1. The Casein Yeast Peptone, catalog no. 39396-500G-F, is meant to be used for research-use only and has a MQ level of 200, meaning that only limited information is available for it and that, as such, it is not meant to be used as a raw material in the manufacturing of edible products. For more information on our M-Clarity program, please visit https://www.sigmaaldrich.com/GB/en/life-science/quality-and-regulatory-management/m-clarity-program.

      Helpful?

Reviews

No rating value

Active Filters