Skip to Content
Merck

Skip To

36970

Bromodichloromethane

amylene stabilized, analytical standard

Synonym(s):

Dichlorobromomethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
BrCHCl2
CAS Number:
Molecular Weight:
163.83
EC Number:
200-856-7
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
1697005
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

quality

amylene stabilized

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.497 (lit.)

bp

87 °C (lit.)

mp

−55 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

ClC(Cl)Br

InChI

1S/CHBrCl2/c2-1(3)4/h1H

InChI key

FMWLUWPQPKEARP-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


Still not finding the right product?

Explore all of our products under Bromodichloromethane


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Minh T Do et al.
Environmental health perspectives, 113(4), 418-424 (2005-04-07)
Chlorination disinfection by-products (CDBPs) are produced during the treatment of water with chlorine to remove bacterial contamination. CDBPs have been associated with an increased risk of bladder cancer. There is also some evidence that they may increase the risk of
Teresa L Leavens et al.
Toxicological sciences : an official journal of the Society of Toxicology, 99(2), 432-445 (2007-07-28)
Exposure to bromodichloromethane (BDCM), one of the most prevalent disinfection byproducts in drinking water, can occur via ingestion of water and by dermal absorption and inhalation during activities such as bathing and showering. The objectives of this research were to
Suvarthi Das et al.
Toxicology and applied pharmacology, 269(3), 297-306 (2013-02-27)
Today's developed world faces a major public health challenge in the rise in the obese population and the increased incidence in fatty liver disease. There is a strong association among diet induced obesity, fatty liver disease and development of nonalcoholic



Global Trade Item Number

SKUGTIN
C3411-1MG04061833485798

Questions

Reviews

No rating value

Active Filters