Skip to Content
Merck
All Photos(1)

Documents

35595

Supelco

PCB No 14

analytical standard

Synonym(s):

3,5-Dichlorobiphenyl, 3,5-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2
CAS Number:
Molecular Weight:
223.10
Beilstein:
1944311
Ballschmiter Number:
14
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1cc(Cl)cc(c1)-c2ccccc2

InChI

1S/C12H8Cl2/c13-11-6-10(7-12(14)8-11)9-4-2-1-3-5-9/h1-8H

InChI key

QHZSDTDMQZPUKC-UHFFFAOYSA-N

General description

Polychlorinated biphenyls (PCBs) are groups of chemical compounds which are persistent in the environment and are considered highly toxic to humans and animals. PCB No 14 may be used for environmental analysis and precise quality control of food and feed.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina P Gritsan et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 4(1), 23-32 (2004-12-24)
The dynamics of singlet 3,5-dichloro-2-biphenylnitrene and the products of its rearrangement were monitored by pico- and nanosecond laser flash photolysis and the results were consistent with the predictions of DFT and ab initio molecular orbital calculations.
J Havel et al.
Applied microbiology and biotechnology, 43(5), 914-919 (1995-10-01)
The influence of different forms of substrate administration (either through the vapour phase or the liquid phase) on growth of two bacterial strains on biphenyl, 2-chlorobiphenyl, and 3,5-dichlorobiphenyl has been investigated. During growth with all three compounds, the availability of
H A J Govers et al.
SAR and QSAR in environmental research, 13(2), 271-280 (2002-06-20)
Enthalpies of transfer at 300 K of various partitioning processes were calculated in order to study the suitability of 3D force fields for the calculation of partitioning constants. A 3D fulvic acid (FA) model of dissolved organic carbon (DOC) was
Metabolism of dichlorobiphenyls by hepatic microsomal cytochrome P-450.
M W Kennedy et al.
Biochemical pharmacology, 30(6), 577-588 (1981-03-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service