Skip to Content
Merck
All Photos(1)

Key Documents

36989

Supelco

PCB standard solution 7

analytical standard, 10 μg/mL in isooctane

Synonym(s):

PCB No 28, 52, 101, 138, 153, 180, 209 solution

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€125.00

€125.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
2 ML
€125.00

About This Item

Ballschmiter Number:
28,52,101,138,153,180,209
UNSPSC Code:
41116107
NACRES:
NA.24

€125.00


Estimated to ship on16 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

Contains 10 ng/μl of:
2,4,4′-PCB; 2,2′,5,5′-PCB; 2,2′,4,5,5′-PCB; 2,2′,3,4,4′,5-PCB; 2,2′,4,4′,5,5′-PCB; 2,2′,3,4,4′,5,5′-PCB; 2,2′,3,3′,4,4′,5,5′,6,6′-PCB

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service