Skip to Content
Merck
All Photos(1)

Key Documents

861291

Supelco

PAH Mix 3

certified reference material, in methylene chloride: methanol (1:1) (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

description

Separate Source

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

in methylene chloride: methanol (1:1) (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CRM47930CRM4890549156
PAH Mix 3 certified reference material, in methylene chloride: methanol (1:1) (varied)

861291

PAH Mix 3

format

multi-component solution

format

multi-component solution

format

multi-component solution

format

multi-component solution

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 1 mL

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene 1000 μg/mL
    Acenaphthylene 2000 μg/mL
    Anthracene 100 μg/mL
    Benz[a]anthracene 100 μg/mL
    Benzo[b]fluoranthene 100 μg/mL
    Benzo[k]fluoranthene 100 μg/mL
    Benzo[ghi]perylene 200 μg/mL
    Benzo[a]pyrene 100 μg/mL
    Chrysene 100 μg/mL
    Dibenz[a,h]anthracene 200 μg/mL
    Fluoranthene 100 μg/mL
    Fluorene 200 μg/mL
    Indeno[1,2,3-cd]pyrene 100 μg/mL
    1-Methylnaphthalene 1000 μg/mL
    2-Methylnaphthalene 1000 μg/mL
    Naphthalene 1000 μg/mL
    Phenanthrene 100 μg/mL
    Pyrene 100 μg/mL
    See All (18)

Signal Word

Danger

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. What is the concentration of this PAH mix 3 (861291)

    1 answer
    1. The concentration is in methylene chloride: methanol (1:1).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service