Skip to Content
Merck
All Photos(2)

Key Documents

34391

Supelco

5-Bromo-5-nitro-1,3-dioxane

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€72.40

€72.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€72.40

About This Item

Empirical Formula (Hill Notation):
C4H6BrNO4
CAS Number:
Molecular Weight:
212.00
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€72.40


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

58-60 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)C1(Br)COCOC1

InChI

1S/C4H6BrNO4/c5-4(6(7)8)1-9-3-10-2-4/h1-3H2

InChI key

XVBRCOKDZVQYAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Bromo-5-nitro-1,3-dioxane is a bromine containing preservative commonly used in cosmetic products.[1]

Application

5-Bromo-5-nitro-1,3-dioxane may be used as an analytical reference standard for the determination of the analyte in rinse-off cosmetics by solid-phase microextraction gas chromatography with microelectron-capture detection method (SPME-GC-μECD).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a solid-phase microextraction gas chromatography with microelectron-capture detection method for the determination of 5-bromo-5-nitro-1, 3-dioxane in rinse-off cosmetics.
Fernandez-Alvarez M, et al.
Journal of Chromatography A, 1217(43), 6634-6639 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service