Skip to Content
Merck

Skip To

32007

Imazalil

PESTANAL®, analytical standard

Synonym(s):

1-[2-(Allyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 77,20

About This Item

Empirical Formula (Hill Notation):
C14H14Cl2N2O
CAS Number:
Molecular Weight:
297.18
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
252-615-0
Beilstein/REAXYS Number:
545683
MDL number:

€ 77,20


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(C(Cn2ccnc2)OCC=C)c(Cl)c1

InChI

1S/C14H14Cl2N2O/c1-2-7-19-14(9-18-6-5-17-10-18)12-4-3-11(15)8-13(12)16/h2-6,8,10,14H,1,7,9H2

InChI key

PZBPKYOVPCNPJY-UHFFFAOYSA-N

General description

Imazalil is a fungicide mostly used to avoid decay during postharvest. It is mostly used for fruits, vegetables and ornamental plants to control fungi growth.[1]

Application

Imazalil has been used as reference standard in direct analysis in real time mass spectrometry (DART-MS) technique for determination of xenobiotics from fruit samples. It may have been used as reference standard in Flow injection photoinduced chemiluminescence method for determination of Imazalil concentration from water samples.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CRM53259Y000013632532
format

neat

format

-

format

neat

format

neat

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

pharmaceutical primary standard

grade

analytical standard

product line

PESTANAL®

product line

TraceCERT®

product line

-

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

application(s)

-

application(s)

pharmaceutical (small molecule)

application(s)

agriculture
environmental


Still not finding the right product?

Explore all of our products under Imazalil


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters